Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGDIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176650.3 | NP_788823.1 | 4 - 5 | 424 | 70 | |
GenBank mRNA | BC103465.1 | - | 4 - 5 | 424 | 70 | |
BT021511.1 | - | 4 - 5 | 405 | 70 | ||
X52689.1 | - | 394 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |