Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177497.2 | NP_803463.1 | 4 - 5 | 494 | 66 | |
GenBank mRNA | AF461050.1 | - | 4 - 5 | 494 | 66 | |
BC111256.1 | - | 5 - 6 | 1314 | 66 | ||
BC120218.1 | - | 5 - 6 | 556 | 66 | ||
BC149572.1 | - | 5 - 6 | 615 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |