Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178320.2 | NP_847890.1 | 2 - 3 | 116 | 76 | |
GenBank mRNA | AB099010.1 | - | 2 - 3 | 116 | 76 | |
AF228021.1 | - | 2 - 3 | 112 | 76 | ||
AY247029.1 | - | 2 - 3 | 116 | 76 | ||
AY911355.1 | - | 2 - 3 | 151 | 76 | ||
BC105173.1 | - | 153 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Recommended Assay Bos taurus Inventoried Probe spans exons |