Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181667.1 | NP_858053.1 | 3 - 4 | 1489 | 80 | |
GenBank mRNA | AF192770.1 | - | 1 - 2 | 94 | 80 | |
AF516915.1 | - | 3 - 4 | 1489 | 80 | ||
BC148059.1 | - | 3 - 4 | 1501 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |