Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181667.1 | NP_858053.1 | 4 - 5 | 1666 | 111 | |
GenBank mRNA | AF192770.1 | - | 2 - 3 | 271 | 111 | |
AF516915.1 | - | 4 - 5 | 1666 | 111 | ||
BC148059.1 | - | 4 - 5 | 1678 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |