Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181667.1 | NP_858053.1 | 5 - 6 | 1946 | 88 | |
GenBank mRNA | AF516915.1 | - | 5 - 6 | 1946 | 88 | |
BC148059.1 | - | 5 - 6 | 1958 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |