Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183085.1 | NP_898908.1 | 4 - 5 | 536 | 87 | |
GenBank mRNA | AY336967.1 | - | 4 - 5 | 536 | 87 | |
BC147981.1 | - | 4 - 5 | 538 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |