Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RETN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183362.1 | NP_899206.1 | 3 - 4 | 320 | 71 | |
GenBank mRNA | AB117718.1 | - | 3 - 4 | 320 | 71 | |
BC148022.1 | - | 3 - 4 | 239 | 71 | ||
JQ353712.1 | - | 3 - 4 | 225 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |