Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POFUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205782.1 | NP_991351.1 | 3 - 4 | 534 | 63 | |
GenBank mRNA | AJ575655.1 | - | 3 - 4 | 534 | 63 | |
AY344582.2 | - | 3 - 4 | 534 | 63 | ||
BC140500.1 | - | 3 - 4 | 529 | 63 | ||
DQ138294.1 | - | 3 - 4 | 534 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |