Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010995.2 | NP_001010995.2 | 5 - 6 | 657 | 81 | |
GenBank mRNA | BC104538.1 | - | 5 - 6 | 657 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |