Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015512.3 | NP_001015512.1 | 1 | 40 | 63 | |
GenBank mRNA | BC102057.1 | - | 1 | 41 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |