Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015631.2 | NP_001015631.1 | 9 - 10 | 1216 | 106 | |
GenBank mRNA | BC104498.1 | - | 9 - 10 | 1216 | 106 | |
BT020831.1 | - | 9 - 10 | 1229 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |