Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024469.3 | NP_001019640.2 | 1 | 62 | 75 | |
GenBank mRNA | AB099048.1 | - | 1 | 46 | 75 | |
AY017376.1 | - | 1 | 80 | 75 | ||
AY911321.1 | - | 2 - 3 | 67 | 75 | ||
BC103427.1 | - | 1 | 63 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |