Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025319.2 | NP_001020490.1 | 4 - 5 | 606 | 123 | |
GenBank mRNA | BC149346.1 | - | 4 - 5 | 716 | 123 | |
DQ078267.1 | - | 4 - 5 | 711 | 123 | ||
DQ085625.1 | - | 4 - 5 | 606 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |