Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025334.2 | NP_001020505.1 | 4 - 5 | 439 | 62 | |
GenBank mRNA | AY911367.1 | - | 4 - 5 | 411 | 62 | |
BC102283.1 | - | 4 - 5 | 439 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |