Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033629.2 | NP_001028801.1 | 10 - 11 | 1437 | 68 | |
GenBank mRNA | BC102131.1 | - | 10 - 11 | 1438 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |