Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GULO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034043.2 | NP_001029215.1 | 3 - 4 | 333 | 62 | |
GenBank mRNA | BC102936.1 | - | 3 - 4 | 334 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |