Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034212.2 | NP_001029384.1 | 3 - 4 | 230 | 70 | |
GenBank mRNA | BC102301.1 | - | 3 - 4 | 232 | 70 | |
BT030680.1 | - | 3 - 4 | 226 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |