Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034212.2 | NP_001029384.1 | 7 - 8 | 638 | 74 | |
GenBank mRNA | BC102301.1 | - | 7 - 8 | 640 | 74 | |
BT030680.1 | - | 7 - 8 | 634 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |