Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034236.2 | NP_001029408.1 | 4 - 5 | 397 | 146 | |
GenBank mRNA | BC102350.1 | - | 4 - 5 | 398 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |