Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034266.2 | NP_001029438.1 | 5 - 6 | 722 | 79 | |
GenBank mRNA | BC102182.1 | - | 5 - 6 | 724 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |