Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034284.2 | NP_001029456.1 | 1 - 2 | 58 | 115 | |
GenBank mRNA | BC102535.1 | - | 1 - 2 | 59 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |