Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034299.1 | NP_001029471.1 | 8 - 9 | 1050 | 78 | |
GenBank mRNA | BC102308.1 | - | 8 - 9 | 1050 | 78 | |
BT021601.1 | - | 8 - 9 | 981 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |