Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034336.1 | NP_001029508.1 | 1 | 41 | 81 | |
GenBank mRNA | BC102967.1 | - | 1 | 41 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |