Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034399.1 | NP_001029571.1 | 3 - 4 | 134 | 82 | |
GenBank mRNA | BC104508.1 | - | 3 - 4 | 134 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |