Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034403.1 | NP_001029575.1 | 2 - 3 | 343 | 79 | |
GenBank mRNA | BC104594.1 | - | 2 - 3 | 343 | 79 | |
BT029797.1 | - | 2 - 3 | 325 | 79 | ||
BT030748.1 | - | 2 - 3 | 331 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |