Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB5IF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034421.2 | NP_001029593.1 | 2 - 3 | 394 | 58 | |
GenBank mRNA | BC102069.1 | - | 2 - 3 | 395 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |