Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034425.1 | NP_001029597.1 | 3 - 4 | 447 | 129 | |
GenBank mRNA | BC103231.1 | - | 3 - 4 | 447 | 129 | |
BT030520.1 | - | 3 - 4 | 449 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |