Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLGRKT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034426.2 | NP_001029598.1 | 2 - 3 | 242 | 136 | |
GenBank mRNA | BC102121.1 | - | 2 - 3 | 243 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |