Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034449.2 | NP_001029621.1 | 7 - 8 | 1055 | 99 | |
GenBank mRNA | BC103363.1 | - | 7 - 8 | 1056 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |