Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034463.2 | NP_001029635.1 | 1 - 2 | 176 | 79 | |
GenBank mRNA | BC102260.1 | - | 1 - 2 | 177 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |