Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034463.2 | NP_001029635.1 | 4 - 5 | 472 | 109 | |
GenBank mRNA | BC102260.1 | - | 4 - 5 | 473 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |