Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034479.1 | NP_001029651.1 | 4 - 5 | 500 | 76 | |
GenBank mRNA | DQ176046.1 | - | 5 - 6 | 602 | 76 | |
DQ176047.1 | - | 4 - 5 | 500 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Inventoried Probe spans exons |