Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034488.2 | NP_001029660.1 | 2 - 3 | 231 | 65 | |
GenBank mRNA | BC102567.1 | - | 2 - 3 | 232 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |