Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034546.2 | NP_001029718.1 | 5 - 6 | 820 | 124 | |
GenBank mRNA | BC104563.1 | - | 5 - 6 | 821 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |