Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034550.2 | NP_001029722.1 | 3 - 4 | 523 | 83 | |
GenBank mRNA | BC102258.1 | - | 3 - 4 | 524 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |