Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034562.1 | NP_001029734.1 | 14 - 15 | 1350 | 89 | |
GenBank mRNA | BC102450.1 | - | 14 - 15 | 1350 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |