Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034596.2 | NP_001029768.1 | 1 - 2 | 84 | 60 | |
GenBank mRNA | BC102554.1 | - | 1 - 2 | 85 | 60 | |
BC151435.1 | - | 1 - 2 | 84 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |