Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034617.1 | NP_001029789.1 | 5 - 6 | 919 | 86 | |
GenBank mRNA | BC102371.1 | - | 5 - 6 | 919 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |