Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034618.2 | NP_001029790.1 | 10 - 11 | 1129 | 88 | |
GenBank mRNA | BC103352.1 | - | 10 - 11 | 1130 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |