Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034628.3 | NP_001029800.1 | 8 - 9 | 1330 | 88 | |
NM_001320319.1 | NP_001307248.1 | 8 - 9 | 1330 | 88 | ||
NR_135204.1 | 7 - 8 | 1154 | 88 | |||
GenBank mRNA | BC102133.1 | - | 8 - 9 | 926 | 88 | |
CO872328.1 | - | 3 - 4 | 253 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |