Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034662.2 | NP_001029834.1 | 3 - 4 | 276 | 122 | |
GenBank mRNA | BC102206.1 | - | 3 - 4 | 277 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |