Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034664.1 | NP_001029836.1 | 2 - 3 | 53 | 80 | |
GenBank mRNA | BC105272.1 | - | 2 - 3 | 53 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |