Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTKN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034681.1 | NP_001029853.1 | 9 - 10 | 1175 | 106 | |
GenBank mRNA | BC102397.1 | - | 9 - 10 | 1175 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |