Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034719.2 | NP_001029891.1 | 2 - 3 | 144 | 102 | |
GenBank mRNA | BC102210.1 | - | 2 - 3 | 146 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |