Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034719.2 | NP_001029891.1 | 6 - 7 | 644 | 60 | |
GenBank mRNA | BC102210.1 | - | 6 - 7 | 646 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |