Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034727.3 | NP_001029899.1 | 4 - 5 | 648 | 75 | |
NM_001271737.1 | NP_001258666.1 | 5 - 6 | 622 | 75 | ||
GenBank mRNA | BC102639.1 | - | 4 - 5 | 622 | 75 | |
EH145309.1 | - | 5 - 6 | 623 | 75 | ||
EH156163.1 | - | 4 - 5 | 442 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |