Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034751.1 | NP_001029923.1 | 2 - 3 | 289 | 63 | |
GenBank mRNA | BC104582.1 | - | 2 - 3 | 289 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |