Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT4H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034792.2 | NP_001029964.1 | 3 - 4 | 262 | 80 | |
GenBank mRNA | BC103340.1 | - | 3 - 4 | 263 | 80 | |
BT025461.1 | - | 3 - 4 | 270 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |