Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LENG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034804.2 | NP_001029976.1 | 2 - 3 | 342 | 79 | |
GenBank mRNA | BC102195.1 | - | 2 - 3 | 343 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |